Invitrogen™

E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%

Catalog number:  G8351ST

 Related applications:

Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%, provides everything you need to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.

The kit includes:
• 18 E-Gel agarose gels with SYBR Safe, 1.2% (Cat. No. G521801)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)

Learn more about the E-Gel Power Snap Electrophoresis System ›

Learn more about precast E-Gel EX agarose gel technology ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: E-Gel™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

• 1 electrophoresis device
• 1 camera device
• 18 E-Gel SYBR Safe 1.2% gels
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter

Store at room temperature.

Documents

Manuals & protocols