Invitrogen™
E-Gel™ Power Snap Plus Electrophoresis Systems, Accessories with Extended Warranty
Catalog number: G9392
Invitrogen™
E-Gel™ Power Snap Plus Electrophoresis Systems, Accessories with Extended Warranty
Catalog number: G9392
The all-in-one Invitrogen E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput E-Gel agarose gel electrophoresis, from DNA sample loading to gel imaging. With dry precast E-Gel agarose gel technology, you can run DNA samples in as little as 10 minutes and observe sample separation in real time. It is the only benchtop system that seamlessly integrates high-throughput DNA sample separation and gel imaging into one workflow.
Catalog Number
G9392
Unit Size
1 package
Includes
Electrophoresis device with integrated camera, E-Gel 96 2% with SYBR Safe, WiFi dongle
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 5,682.41
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 96 2% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
Catalog Number
Unit Size
1 device
Includes
Electrophoresis device only
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 2,606.28
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device only
Mode of SeparationElectrophoresis
Product TypeElectrophoresis Device
Quantity1 Device
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 device
Catalog Number
Unit Size
1 camera
Includes
Camera only
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 2,972.70
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
For Use With (Equipment)E-Gel™ Power Snap Plus Electrophoresis Device
IncludesCamera only
Product TypeCamera
Quantity1 Camera
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 camera
Catalog Number
Unit Size
1 system
Includes
Electrophoresis device with integrated camera, WiFi dongle
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 5,608.91
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeElectrophoresis System
Quantity1 System
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 system
Catalog Number
Unit Size
1 package
Includes
Electrophoresis device with integrated camera, E-Gel 48 1% with SYBR Safe, WiFi dongle
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 5,680.91
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 48 1% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
Catalog Number
Unit Size
1 package
Includes
Electrophoresis device with integrated camera, E-Gel 48 2% with SYBR Safe, WiFi dongle
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 5,681.66
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 48 2% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
Catalog Number
Unit Size
1 package
Includes
Electrophoresis device with integrated camera, E-Gel 96 1% with SYBR Safe, WiFi dongle
Warranty
1-year rapid exchange extended warranty included
Price (USD)
Price: 5,674.91
Your Price:
Full specifications
Quantity
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 96 1% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeIncludesWarrantyPrice (USD)Quantity
G9392Full specifications
1 packageElectrophoresis device with integrated camera, E-Gel 96 2% with SYBR Safe, WiFi dongle1-year rapid exchange extended warranty included
Price: 5,682.41
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 96 2% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
G9101Full specifications
1 deviceElectrophoresis device only1-year rapid exchange extended warranty included
Price: 2,606.28
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device only
Mode of SeparationElectrophoresis
Product TypeElectrophoresis Device
Quantity1 Device
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 device
G9201Full specifications
1 cameraCamera only1-year rapid exchange extended warranty included
Price: 2,972.70
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
For Use With (Equipment)E-Gel™ Power Snap Plus Electrophoresis Device
IncludesCamera only
Product TypeCamera
Quantity1 Camera
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 camera
G9301Full specifications
1 systemElectrophoresis device with integrated camera, WiFi dongle1-year rapid exchange extended warranty included
Price: 5,608.91
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeElectrophoresis System
Quantity1 System
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 system
G9341Full specifications
1 packageElectrophoresis device with integrated camera, E-Gel 48 1% with SYBR Safe, WiFi dongle1-year rapid exchange extended warranty included
Price: 5,680.91
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 48 1% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
G9342Full specifications
1 packageElectrophoresis device with integrated camera, E-Gel 48 2% with SYBR Safe, WiFi dongle1-year rapid exchange extended warranty included
Price: 5,681.66
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 48 2% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
G9391Full specifications
1 packageElectrophoresis device with integrated camera, E-Gel 96 1% with SYBR Safe, WiFi dongle1-year rapid exchange extended warranty included
Price: 5,674.91
Your Price:
ColorWhite
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
Gel Percentage0.8 to 4%
Gel TypeE-Gel, E-PAGE
IncludesElectrophoresis device with integrated camera, E-Gel 96 1% with SYBR Safe, WiFi dongle
Mode of SeparationElectrophoresis
No. of Wells11 to 96-wells
Product TypeStarter Kit
Quantity1 Kit
Sample TypeProtein, DNA
Separation Range10 bp to 15 kb
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
Unit Size1 package
Showing 7 of 7
Features of the E-Gel Power Snap Plus Electrophoresis system include:
• Streamline electrophoresis and image capture with an all-in-one system
• Compatible with 11- to 96-well E-Gel agarose gels
• View and capture DNA samples in real-time
• Access your images anywhere using the built-in Thermo Fisher Connect and network compatibility
Small, fast, and powerful
The all-in-one E-Gel Power Snap Plus Electrophoresis System provides everything you need for DNA gel electrophoresis and gel visualization in a small lightweight benchtop system. The system includes an electrophoresis power supply, blue-light transilluminator, amber filter, and adapter for 11- to 22-well E-Gel agarose gels. This enables gel separation and real-time sample tracking on E-Gel agarose gels pre-stained with SYBR Safe or SYBR Gold II DNA stains. The device arrives with pre-programmed protocols for each type of compatible E-Gel agarose gel.
Instant capture and seamless integration
Capture high-resolution E-Gel agarose gel images with the included easy-to-use E-Gel Power Snap Plus Camera. The camera is directly docked to the E-Gel Power Snap Electrophoresis Device, requiring no external power supply or connection to a desktop computer. The E-Gel Power Snap Plus Camera is also compatible with the included WiFi dongle and has an ethernet port, so you can export your experimental data to the cloud or internal servers for maximum convenience.
Unsurpassed convenience of dry precast E-Gel agarose gel
Run DNA samples in as little as 10 minutes with dry, precast, pre-stained E-Gel agarose gels. The E-Gel dry precast agarose gel technology provides speed and convenience, eliminating any need for gel preparation or staining. E-Gel agarose gels are available in a variety of gel percentage and a choice of two DNA stains. The E-Gel Power Snap Plus Electrophoresis Device is compatible with high-throughput E-Gel 48 and E-Gel 96 gels with SYBR Safe*. A provided adapter extends compatibility to low-throughput E-Gel precast agarose gels, including E-Gel EX, E-Gel SYBR Safe, E-Gel CloneWell II, and E-Gel SizeSelect II gels*.
Safer workflows
The blue light emitted by the E-Gel Power Snap Plus Electrophoresis Device transilluminator ensures maximum possible safety to the user and the samples. A combination of blue light and amber filter also offers real-time viewing of the migration of your samples. The E-Gel Power Snap Plus system is a perfect choice for routine PCR fragment analysis, genotyping, and other applications requiring high-throughput. The compatibility with E-Gel CloneWell II gels also makes it a great choice for a safer cloning workflow with increased efficiency.
Made for convenience
The integrated E-Gel Power Snap Plus Camera (available separately as G9201) touch-screen and on-board software provide an intuitive interface for quick gel image capture and editing. The device also offers built in E-Gel 96 deconvolution into several more readable formats. You can store images in the internal memory of the Power Snap Plus system, on our Thermo Fisher Connect cloud-based platform, or on local network drive to further optimize your workflow.
*The E-Gel Power Snap Plus Electrophoresis System also supports ethidium bromide-stained E-Gel high- and low- throughput agarose gels. The ethidium bromide-stained gels are not suitable for imaging with the E-Gel Power Snap Camera or real-time view.
For Research Use Only.
Figures