Fixation Medium (Medium A)
Invitrogen™

Fixation Medium (Medium A)

Fixation Medium (Medium A) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
GAS001S100100 mL
GAS001S55 mL
Catalog number GAS001S100
Price (USD)
1,236.00
Each
Quantity:
100 mL
Price (USD)
1,236.00
Each
Fixation Medium (Medium A) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.

The FIX & PERM™ Cell Permeabilization Kit is:

• Flexible—compatible with multiple sample types and cellular antigens
• Robust—scatter pattern of cells unaffected
• Efficient—simultaneous detection of intracellular and surface markers

View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).

The Fixation Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.

How It Works
Cells are incubated for 15 minutes at room temperature with the Fixation Medium and washed. Cells can then be incubated with Permeabilization Medium (Medium B) (Cat. No. GAS002S100) and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGray
FormLiquid
Quantity100 mL
Reagent TypeFixation Reagent
Shipping ConditionRoom Temperature
Storage BufferFormaldehyde
Volume (Metric)100 mL
For Use With (Application)Flow Cytometry
Product LineFIX and PERM
Product TypeFixation Medium
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

The FIX & PERM Cell Permeabilization Kit protocol does not say what volume my cells should be in when they are added to the tube. What should I do?

Your cells should be in approximately 100 µL volume. One way to do this is to aliquot the correct number of cells (10E6) and then spin them down and decant the supernatant. The residual volume should be approximately 100 µL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the FIX & PERM Cell Permeabilization Kit to access nuclear epitopes?

Yes. Generally, you need to increase the incubation time in the permeabilization buffer (Reagent B). Here are a couple of references:
-Millard I, Degrave E, Philippe M et al. (1998) Detection of intracellular antigens by flow cytometry: comparison of two chemical methods and microwave heating. Clin Chem 44:2320-2330(http://www.ncbi.nlm.nih.gov/pubmed/9799760).
-Parker JE, Mufti GJ, Rasool F et al. (2000) The role of apoptosis, proliferation, and the Bcl-2-related proteins in the myelodysplastic syndromes and acute myeloid leukemia secondary to MDS. Blood 96:3932-3938(http://www.ncbi.nlm.nih.gov/pubmed/11090080).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

The FIX & PERM Cell Permeabilization Kit is used to prepare cells for flow cytometry; can I use it for immunohistochemistry?

Yes. Here are a couple of references:
Haranaga S, Yamaguchi H, Friedman H et al.(2001) Chlamydia pneumoniae infects and multiplies in lymphocytes In vitro. Infect Immun 69:7753-7759 (http://www.ncbi.nlm.nih.gov/pubmed/11705957).

Al-Mufti R, Hambley H, Albaiges G et al. (2000) Increased fetal erythroblasts in women who subsequently develop pre-eclampsia. Hum Reprod 15:1624-1628 (http://www.ncbi.nlm.nih.gov/pubmed/10875878).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Does the FIX & PERM Cell Permeabilization Kit (Cat. Nos. GAS003 and GAS004) lyse red blood cells?

Yes. The FIX & PERM Cell Permeabilization Kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics. It has been validated for use in cell lines, whole blood, and bone marrow. In addition fixing and permeabilizing cells, it lyses red blood cells. It is labeled as a general purpose reagent (GPR) and can be used in clinical settings, but it is also used widely in research labs.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.