Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Actual product may vary
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Thermo Scientific™
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
GCF400Condensation Flask
SCT120Chemical Trap
DTK120RChemical Trap Kit
DC120Disposable Cartridge
DC120AAcid Neutralization Cartridge
ANC100Acid Neutralizing Compound
UTP-TEF0.75
also known as UTP-TEF 0.75
Tubing Pack
ANS121Ammonia Neutralizing Solution
ANT100Post Trap Assembly
MF190Mist Filter Replacement Cartridge
CC120DX
also known as CC120/DX
Cart
Catalog number GCF400
Price (USD)
684.00
Online Exclusive
760.00
Save 76.00 (10%)
Each
Type:
Condensation Flask
Price (USD)
684.00
Online Exclusive
760.00
Save 76.00 (10%)
Each
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
Specifications
DescriptionGlass condensation flask
For Use With (Equipment)SPD1030 and SPD2030 DNA SpeedVac™ Concentrator
TypeCondensation Flask
Unit SizeEach