Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Thermo Scientific™

Orion™ Combination pH Electrodes

Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFilling Solution TypeJunction
GD9106BNWPGel-filledDouble
GD9156BNWPLiquid-filledDouble
GS9106BNWPGel-filledSingle
Catalog number GD9106BNWP
Price (USD)
223.20
Online Exclusive
248.00
Save 24.80 (10%)
Each
Filling Solution Type:
Gel-filled
Junction:
Double
Price (USD)
223.20
Online Exclusive
248.00
Save 24.80 (10%)
Each
Thermo Scientific™ Orion™ Green Electrodes are the first lab electrode to contain no lead, mercury or other hazardous substances and are completely ROHS compliant. GD9156BNWP is a refillable electrode that provides reliable and accurate measurements of routine samples but are best suited for treated effluent, distilled water and wastewater. GD9106BNWP and GS9106BNWP contain a low-maintenance gel that is best suited for field or plant use and educational institutes, where minimal maintenance is required.
 
Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples. While Orion™ Single Junction Electrodes provide a reliable and accurate option for routine pH measurement of typical samples.
 
Solutions required 
Electrode fill solution for GD9156BNWP
910008     3M KCl filling solution, 5 x 60 mL bottles 
 
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with GD9156BNWP
910008     3M KCl filling solution 60mL 1 bottle
 
Warranty period
GD9156BNWP: 12 months from date of purchase
GD9106BNWP and GS9106BNWP: 6 months from date of purchase
Specifications
DescriptionLow-maintenance gel-filled
Filling Solution TypeGel-filled
For Use With (Equipment)Orion pH Meter
Length (English) Cable39.3 in.
Length (Metric) Cable1 m
Connector TypesBNC (Waterproof), BNC-to-USS adapter is required with this product as a onetime purchase. Part Number: 13-641-890
JunctionDouble
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 90°C
TypeGreen Combination pH Electrode, gel-filled
pH Range0 to 14
Unit SizeEach