Product Image
Applied Biosystems™
dC(Bz) Phosphoramidite, 2.0 g.
This dC(Bz) Phosphoramidite is used for nucleic acid synthesis on the Expedite™ 8905, 8909, and 8909 with MOSS upgrade DNA synthesizers.
Have Questions?
Catalog NumberQuantity
GEN0660122 g
Catalog number GEN066012
Price (HKD)
-
Quantity:
2 g
This dC(Bz) Phosphoramidite is used for nucleic acid synthesis on the Expedite™ 8905, 8909, and 8909 with MOSS upgrade DNA synthesizers. Our β-cyanoethyl phosphoramidites guarantee you high-quality nucleic acid synthesis.
  • Proprietary manufacturing processes guarantee high purity products
  • Expedite™ system fast deprotecting amidites offer fast deprotection for primer-length oligonucleotides, shortening overall production time and costs
  • Fast deprotecting amidites and standard deprotecting amidites are use-tested and optimized for use on the Expedite™ System for high-quality nucleic acid synthesis
  • Phosphoramidite kits simplify ordering, saving you time and money
High purity
Our phosphoramidites benefit from proprietary manufacturing processes that yield products of high purity. In synthesis they perform at correspondingly high levels, producing pure, high quality DNA at all scales. We guarantee that 5'-dimethoxytrityl nucleoside β-cyanoethyl phosphoramidites, monomer intermediates for licensed β-cyanoethyl phosphoramidite chemistry, will produce DNA that is at least 99.8% chemically authentic when used with recommended synthesis cycles.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)3900 DNA Synthesizer
FormatPowder
Nucleic Acid TypeDNA
Nucleoside TypeDeoxycytidine (dC)
Product TypedC(Bz) CEP
Quantity2 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit Size2 g
Contents & Storage
Store in freezer (-5 to -30°C).