Human BCRP Vesicles are insect derived purified plasma membranes with inserted BCRP transport protein (Breast Cancer Resistance Protein, ABCG2). Our자세히 알아보기
Have Questions?
카탈로그 번호
수량
GM0008
0.5 mL
카탈로그 번호 GM0008
제품 가격(KRW)
1,890,000
온라인 행사
Ends: 31-Mar-2026
2,099,000
할인액 209,000 (10%)
Each
수량:
0.5 mL
제품 가격(KRW)
1,890,000
온라인 행사
Ends: 31-Mar-2026
2,099,000
할인액 209,000 (10%)
Each
Human BCRP Vesicles are insect derived purified plasma membranes with inserted BCRP transport protein (Breast Cancer Resistance Protein, ABCG2). Our Human BCRP Vesicles can be used to evaluate test compounds and drug interactions with BCRP in in vitro assays. BSEP is an ATP-binding cassette (ABC) transporter associated with multidrug resistance, which carries various anti-cancer drugs such as topotecan, methotrexate and mitoxantrone.
Use Human BCRP Vesicles to:
• Investigate the transporter interactions of your drug candidates • Assess potential for transporter-mediated drug-drug interactions • Obtain high quality results with a large signal to noise ratios
Concept Behind ABC Transporter Vesicles ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.
Clear and Reliable Results Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.
Transporter Vesicle Details Human BCRP Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing BCRP. It is highly expressed in tumors like lung cancer and also in small intestinal epithelial cells, hepatic parenchymal cells, proximal renal tubule cells, and cerebrovascular endothelial cells. It is thought to be an important determinant of drug disposition, through its role in processes such as absorption at the small intestine, excretion into bile and urine, and transfer across the blood-brain barrier to the CNS.
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
사양
형식Vial
제품 유형Human Vesicle
순도Research Grade
수량0.5 mL
배송 조건Dry Ice
숙주 종Human
물리적 형태Frozen
Unit SizeEach
구성 및 보관
Store in ultracold freezer (-60 to -85°C) and protect from light.
자주 묻는 질문(FAQ)
What is the supplied quantity of ATP-binding cassette (ABC) Transporter Vesicles?
The quantity supplied is 2.5 mg total protein/500 µL per vial. The protein concentration is 5 mg/mL. When our recommended protocol is followed, 50 vesicular transport assays or 125 ATPase assays can be performed with one vial of ABC Transporter Vesicles product.
What are ATP-binding cassette (ABC) Transporter Vesicles?
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, ATP-binding cassette (ABC) Transporter Vesicles are inside-out vesicles designed for use in vesicular transport assays. They allow the investigation of possible interactions between specific ABC transporters and test compounds. ABC Transporter Vesicles provide high levels of transporter activity with low background, giving a clear signal if the test compound is a substrate or inhibitor of a specific efflux transporter.
While ABC transporters typically mediate the efflux of substrates from cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for the test compound by determining the amount incorporated into the vesicles.
Note: One vial of vesicle product can perform 50 vesicular transport assays.