Product Image
Gibco™
BSEP Vesicular Transport Assay Reagent Set
The BSEP Vesicular Transport Assay Reagent Set is a ready-to-use kit containing all reagents needed for vesicular transport assay except深入閱讀
Have Questions?
產品號碼Quantity
GM3001100 Assays
產品號碼 GM3001
價格 (TWD)
-
Quantity:
100 Assays
The BSEP Vesicular Transport Assay Reagent Set is a ready-to-use kit containing all reagents needed for vesicular transport assay except for the vesicles themselves. The reagent set can be used for vesicular transport assay using radioisotope-labeled or non-labeled compound. This kit has enough material to perform 100 vesicular transport assays.
Use The BSEP Vesicular Transport Assay Reagent Set to:
• Investigate the transporter interactions of your drug candidates
• Assess potential for transporter-mediated drug-drug interactions
• Obtain high quality results with a large signal to noise ratios
Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate’s substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.
Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.
Kit Compatibility
Our BSEP Vesicular Transport Assay Reagent Set is designed for use with BSEP Vesicles. The 100 vesicular transport assays that can be performed with this kit will require two BSEP Vesicle products
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
規格
PackagingBottle
Product TypeAssay Reagent Set
Quantity100 Assays
Shipping ConditionDry Ice
FormFrozen
Unit SizeEach
內容物與存放
Store at -20°C;.