Ion ReproSeq™ PGT-A GX5 Kit, 192 samples - FAQs
Ion ReproSeq™ PGT-A GX5 Kit, 192 samples - FAQs
View additional product information for Ion ReproSeq™ PGT-A GX5 Kit - FAQs (GX34902, GX34903)
8 product FAQs found
With the Ion ReproSeq PGT-A GX5 Kit, what is the turnaround time (sample to test results) for PGT-A + SNP QC samples?
The turnaround time for PGT-A + SNP QC samples is estimated to be 21 hr.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
With the Ion ReproSeq PGT-A GX5 Kit, what is the turnaround time (sample to test results) for SNP QC-only samples?
The turnaround time for SNP QC-only samples is estimated to be 18 hr (48 samples).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
With the Ion ReproSeq PGT-A GX5 Kit, what is the turnaround time (sample to test results) for PGT-A-only samples?
The turnaround time for PGT-A-only samples is estimated to be 10 hr 30 min (48 samples) and 19 hr 45 min (192 samples).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What version of Genexus Software is needed to run the Ion ReproSeq PGT-A GX5 Kit?
Genexus Software version 6.8.0 is needed to run the Ion ReproSeq PGT-A GX5 Kit.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
With the Ion ReproSeq PGT-A GX5 Kit, how many reads per sample can I expect for 48 libraries/lane?
48 libraries per lane yields >200,000 reads for most libraries for the Ion ReproSeq PGT-A GX5 Kit.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
With the Ion ReproSeq PGT-A GX5 Kit, how many samples can I run on each chip?
The Ion ReproSeq PGT-A GX5 Kit can run 192 samples/chip (48 samples/lane).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Are the Ion Xpress and IonCode barcodes compatible with the Ion ReproSeq PGT-A GX5 Kit?
Yes, the Ion ReproSeq PGT-A GX5 Kit is compatible with the IonXpress 1-96 and IonCode 1-384 (IonCode_0101-0496) barcodes.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What new barcodes are used for the Ion ReproSeq PGT-A GX5 Kit?
The new barcodes in the Ion ReproSeq PGT-A GX5 Kit are:
- Ion SingleSeq GX 101-196 Kit (Cat. No. A50250)
- Ion SingleSeq GX 201-296 Kit (Cat. No. A50251)
- Ion SingleSeq GX 301-396 Kit (Cat. No. A50252)
- Ion SingleSeq GX 401-496 Kit (Cat. No. A50253)
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.