Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR, Fisher Chemical™
Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR, Fisher Chemical™
Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR, Fisher Chemical™
Fisher Chemical™

Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR, Fisher Chemical™

Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
2.5 L
產品號碼 H-1550-15
亦稱為 H/1550/15
價格 (TWD)
-
Quantity:
1 L
Request bulk or custom format
規格
Boiling Point100°C
CAS7722-84-1
ChEBICHEBI:16240
ColorColorless
Formula Weight34.0128g/mol
檢視更多
Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR