CD19 Monoclonal Antibody (HIB19), NovaFluor™ Blue 510, eBioscience™ - FAQs
CD19 Monoclonal Antibody (HIB19), NovaFluor™ Blue 510, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (HIB19), NovaFluor™ Blue 510, eBioscience™ - FAQs (H004T02B01-A, H004T03B01-A)
1 product FAQs found
What fixation methods are compatible with NovaFluor dyes?
NovaFluor conjugates have been tested to be stable for up to 14 days on cells fixed with 1% formalin, 2% paraformaldehyde, eBioscience IC Fixation Buffer (Cat. No. 00-8222-49), or eBioscience Foxp3 / Transcription Factor Staining Buffer Set (Cat. No. 00-5523-00) when stored at 2-8 degrees C in the dark.
NovaFluor dyes can also be used as a surface stain prior to methanol fixation.