Depend on the heavy-duty, split-phase induction motor in Thermo Scientific™ Barnstead™ RO Distribution Pumps, for use with pure water storage tanks.
Have Questions?
Change view
Catalog Number
Horsepower
Phase
Electrical Requirements
H1112
1/3hp
1
115/230 V 50/60 Hz
H1122
1/2hp
1
115/230 V 50/60 Hz
H1132
1hp
1
115/230 V 50/60 Hz
H1142
1/2-1hp
3
230/460 V 50/60 Hz
4 Options
Catalog number H1112
Price (MXN)
-
Horsepower:
1/3hp
Phase:
1
Electrical Requirements:
115/230 V 50/60 Hz
Depend on the heavy-duty, split-phase induction motor in Thermo Scientific™ Barnstead™ RO (Reverse Osmosis) Distribution Pumps. Distribution pumps are used with purified water storage tanks when it is necessary to keep the pure water under pressure for delivery to locations above tank level or at greater than gravity flow pressure.
Includes:
Plastic pump bypass with restrictive orifice
Low-level cutoff switch
Specifications
Description1/3 HP motor
Electrical Requirements115/230 V 50/60 Hz
Frequency50/60 Hz
Horsepower1/3hp
Phase1
Voltage115/230 V
For Use With (Equipment)Purified Water Storage Tanks