HybriSlip™ Hybridization Cover, 40 mm x 22 mm, RNase free
HybriSlip hybridization covers are nuclease free, ready to use and designed specifically for in situ hybridization. These hydrophobic coverslips doRead more
Have Questions?
Catalog Number
Quantity
H18201
also known as H-18201
1 set
Catalog number H18201
also known as H-18201
Price (USD)
271.00
Each
Quantity:
1 set
Price (USD)
271.00
Each
HybriSlip hybridization covers are nuclease free, ready to use and designed specifically for in situ hybridization. These hydrophobic coverslips do not require lengthy preparation or blocking procedures to prevent probe trapping or binding. HybriSlip covers are heat resistant and do not curl, even at high temperatures. They are ideal for denaturation steps or in situ PCR incubations.[The PCR process is covered by patents owned by Hoffmann-LaRoche, Inc. Purchase of these products does not convey a license under these patents.]
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions40 x 22 mm
Quantity1 set
Shipping ConditionRoom Temperature
Product LineHybriSlip
TypeHybridization Cover
Unit SizeEach
Contents & Storage
Store at room temperature.
Citations & References (2)
Citations & References
Abstract
Cytogenetic evidence for asexual evolution of bdelloid rotifers.
Authors:Mark Welch JL, Mark Welch DB, Meselson M
Journal:Proc Natl Acad Sci U S A
PubMed ID:14747655
DNA sequencing has shown individual bdelloid rotifer genomes to contain two or more diverged copies of every gene examined and has revealed no closely similar copies. These and other findings are consistent with long-term asexual evolution of bdelloids. It is not entirely ruled out, however, that bdelloid genomes consist of ... More
Analysis of gene expression by in situ hybridization on adult zebrafish brain sections.
Authors:Kühn E, Köster RW,
Journal:CSH Protoc
PubMed ID:20150145
Whole-mount in situ hybridization is a powerful method for the analysis of gene expression in zebrafish. However, the thickness of adult brain tissue often hampers penetration of reagents, making the direct application of protocols normally used for embryonic stages on adult tissues difficult. This protocol presents a detailed procedure for ... More