6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals
6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals
6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H25825.ME
also known as H25825-ME
500 mg
Catalog number H25825.ME
also known as H25825-ME
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS1651-29-2
IUPAC Name6-chloro-2-fluoro-7H-purine
Molecular FormulaC5H2ClFN4
InChI KeyUNRIYCIDCQDGQE-UHFFFAOYSA-N
SMILESFC1=NC(Cl)=C2NC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, crystals, crystalline powder and/or chunks
Water Content (Karl Fischer Titration)≤2.0%
Assay (Silylated GC)≥96.0%
Melting Pointca. 150-160?C dec.
Appearance (Color)White to cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only