Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).
Solubility
Soluble in DMSO, and methanol.
Notes
Stor
RUO – Research Use Only
General References:
- Gokhan Adas,; Soykan Arikan,; Ozgur Kemik,; Ali Oner,; Nilgun Sahip,;Oguzhan Karatepe. Use of albendazole sulfoxide, albendazole sulfone, and combined solutions as scolicidal agents on hydatid cysts (in vitro study). World J Gastroenterol. 2009, 15 (1), 112-116.
- Katrin Ingold,; Peter Bigler,; Wolfgang Thormann,; Tania Cavaliero,; Bruno Gottstein,; Andrew Hemphill. Efficacies of Albendazole Sulfoxide and Albendazole Sulfone against In Vitro-Cultivated Echinococcus multilocularis Metacestodes. Antimicrob. Agents Chemother. 1999, 43(5), 1052-1061.