(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%
(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%
(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%
(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%
Thermo Scientific Chemicals

(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%

CAS: 31284-89-6 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H25953.03
also known as H25953-03
Price (EUR)
64,30
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
64,30
Each
Add to cart
Chemical Identifiers
CAS31284-89-6
IUPAC Name2-hydroxy-2-(4-methylphenyl)acetic acid
Molecular FormulaC9H10O3
InChI KeySFGURAWGCAPHON-UHFFFAOYNA-N
SMILESCC1=CC=C(C=C1)C(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Enantiomeric excess≥ 97%
Appearance (Color)White
Formcrystals or powder
Assay (Aqueous acid-base Titration)> 97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only