8-Bromoquinoline, 98%, Thermo Scientific Chemicals
8-Bromoquinoline, 98%, Thermo Scientific Chemicals
8-Bromoquinoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromoquinoline, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H25961.03
also known as H25961-03
1 g
Catalog number H25961.03
also known as H25961-03
Price (EUR)
123,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
123,00
Each
Chemical Identifiers
CAS16567-18-3
IUPAC Name8-bromoquinoline
Molecular FormulaC9H6BrN
InChI KeyPIWNKSHCLTZKSZ-UHFFFAOYSA-N
SMILESBrC1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.6700-1.6745 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only