Triphenylsulfonium chloride, 94%
Triphenylsulfonium chloride, 94%
Triphenylsulfonium chloride, 94%
Triphenylsulfonium chloride, 94%
Thermo Scientific Chemicals

Triphenylsulfonium chloride, 94%

CAS: 4270-70-6 | C18H15ClS | 298.828 g/mol
Have Questions?
Catalog NumberQuantity
H26059.03
also known as H26059-03
1 g
Catalog number H26059.03
also known as H26059-03
Price (EUR)
89,50
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
89,50
Each
Add to cart
Chemical Identifiers
CAS4270-70-6
IUPAC Nametriphenylsulfanium chloride
Molecular FormulaC18H15ClS
InChI KeyZFEAYIKULRXTAR-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or yellow
FormPowder
Proton NMRConforms to structure
Assay (Titration ex Chloride)>92.5 to <107.5%
Triphenylsulfonium chloride is used as a key intermediate that is widely used in the new electronic chemicals such as semiconductor, LCD and TFT.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylsulfonium chloride is used as a key intermediate that is widely used in the new electronic chemicals such as semiconductor, LCD and TFT.

Solubility
Soluble in water.

Notes
Store in cool pla
RUO – Research Use Only

General References:

  1. Katsutoshi Ohkubo; Tokio Yamabe; Kenichi Fukui. A Study of the Low-temperature Oxidation of Cumene in the Presence of Triphenylsulfonium Chloride. Propagation and Termination Steps. Bulletin of the Chemical Society of Japan. 1969, 42 (8), 2220-2227.
  2. C. G. Overberger; George Walter Halek. Monomers and Polymers, A Synthesis of Vinyl Cyclopropane and Dicyclopropyl. J. Org. Chem. 1963, 28 (3), 867-868.