2-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
2-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
2-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals

2-Bromo-5-(trifluoromethyl)benzaldehyde, 97%

CAS: 102684-91-3 | C8H4BrF3O | 253.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26109.MD
also known as H26109-MD
Price (EUR)
26,50
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
26,50
Each
Chemical Identifiers
CAS102684-91-3
IUPAC Name2-bromo-5-(trifluoromethyl)benzaldehyde
Molecular FormulaC8H4BrF3O
InChI KeyCSOBJYGHQOLWOD-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(C=O)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5090-1.5130 @ 20?C
Free acid (titration)<3.5%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only