4-n-Hexylaniline, 98%
4-n-Hexylaniline, 98%
4-n-Hexylaniline, 98%
Thermo Scientific Chemicals

4-n-Hexylaniline, 98%

CAS: 33228-45-4 | C12H19N | 177.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26297.14
also known as H26297-14
Price (EUR)
201,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
201,00
Each
Chemical Identifiers
CAS33228-45-4
IUPAC Name4-hexylaniline
Molecular FormulaC12H19N
InChI KeyOVEMTTZEBOCJDV-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to pale yellow to yellow to orange
Identification (FTIR)Conforms
Refractive Index1.5225-1.5265 @ 20?C
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
RUO – Research Use Only

General References:

  1. Zhengjin Jiang et. al. Mixed-mode reversed-phase and ion-exchange monolithic columns for micro-HPLC.. Journal of Separation Science. 2008 , 31 (15) ,2774-2783.
  2. Lioubov I Brueggemann.; Jack M Sullivan.HEK293S cells have functional retinoid processing machinery.. Journal of General Physiology. 2002, 119 (6), 593-612.