Agarose LM, low melt, for recovery of samples after separation
Agarose LM, low melt, for recovery of samples after separation
Agarose LM, low melt, for recovery of samples after separation
Thermo Scientific Chemicals

Agarose LM, low melt, for recovery of samples after separation

Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. | CAS: 9012-36-6 | C24H38O19 | 630.55 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
產品號碼 H26417.22
亦稱為 H26417-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Clarity@1.5% =< 4NTU
CommentEEO =< 0.12
CommentSulphate =< 0.12%
CommentGel strength @1.5% >= 500g/cm²
檢視更多
Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. Ideal for analysis and recovery of DNA and RNA for routine applications. Ideal for a variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays. Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only