p-Sexiphenyl, 95%
p-Sexiphenyl, 95%
p-Sexiphenyl, 95%
Thermo Scientific Chemicals

p-Sexiphenyl, 95%

CAS: 4499-83-6 | C36H26 | 458.604 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26586.03
also known as H26586-03
Price (USD)
460.65
Special offer
Online exclusive
Ends: 30-Jun-2026
542.00
Save 81.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
460.65
Special offer
Online exclusive
Ends: 30-Jun-2026
542.00
Save 81.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS4499-83-6
IUPAC Name4,4'-bis({[1,1'-biphenyl]-4-yl})-1,1'-biphenyl
Molecular FormulaC36H26
InChI KeyZEMDSNVUUOCIED-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥94.0% (based on elemental C)
Elemental AnalysisH: 5.37-5.82% (Theory: 5.71%)
Appearance (Color)White to cream or pale yellow
Elemental AnalysisC: 88.63-96.18% (Theory: 94.29%)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only