Thermo Scientific Chemicals

Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: H26896.14
25 g, Each
Thermo Scientific Chemicals

Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: H26896.14
25 g, Each
Quantity
Catalog number: H26896.14
also known as H26896-14
Price (USD)

Chemical Identifiers

CAS
1067-71-6
IUPAC Name
diethyl (2-oxopropyl)phosphonate
Molecular Formula
C7H15O4P
InChI Key
RSAFKRSMGOSHRK-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC(C)=O)OCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4315-1.4365 @ 20?C

Description

Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text