3-Bromocatecol, 95 %, Thermo Scientific Chemicals
3-Bromocatecol, 95 %, Thermo Scientific Chemicals
3-Bromocatecol, 95 %, Thermo Scientific Chemicals
3-Bromocatecol, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromocatecol, 95 %, Thermo Scientific Chemicals

CAS: 14381-51-2 | C6H5BrO2 | 189.008 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 mg
1 g
Número de catálogo H26925.03
también denominado H26925-03
Precio (USD)
-
Cantidad:
1 g
Pedido a granel o personalizado
Identificadores químicos
CAS14381-51-2
IUPAC Name3-bromobenzene-1,2-diol
Molecular FormulaC6H5BrO2
InChI KeyJPBDMIWPTFDFEU-UHFFFAOYSA-N
SMILESOC1=CC=CC(Br)=C1O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to cream or pale yellow
Assay (GC)≥94.0%
Melting Point (clear melt)36-45?C
Water Content (Karl Fischer Titration)<2.0%
FormCrystalline or lumpy powder or crystals
3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El 3-bromocatecol se utiliza como fármaco, intermedio agroquímico e intermedio farmacéutico.

Solubilidad
Soluble en disolventes orgánicos.

Notas
Almacenar en un lugar fresco. Mantener el recipiente bien cerrado en un lugar seco y bien ventilado. Estable en las condiciones de almacenamiento recomendadas. Mantener alejados los agentes oxidantes fuertes.
RUO – Research Use Only

General References:

  1. T VidaráHansen. Direct biocatalytic synthesis of functionalized catechols: a green alternative to traditional methods with high effective mass yield. Green Chemistry. 20002 (6), 263-265.
  2. UM Reinscheid.; MP Bauer.; R Müller. Biotransformation of halophenols by a thermophilic Bacillus sp. Biodegradation. 19967 (6), 455-461.