Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%
Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%
Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%
Thermo Scientific Chemicals

Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%

CAS: 3939-01-3 | C7H3F2N | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H26930.14
also known as H26930-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3939-01-3
IUPAC Namehydrogen methyl 1-benzyl-4-oxopiperidine-3-carboxylate chloride
Molecular FormulaC14H18ClNO3
InChI KeyBRADBAOVPACOQQ-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C1CN(CC2=CC=CC=C2)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Water Content (Karl Fischer Titration)≤2.5%
Appearance (Color)White to pale cream
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only