Ammonia, 2M in methanol
Ammonia, 2M in methanol
Ammonia, 2M in methanol
Thermo Scientific Chemicals

Ammonia, 2M in methanol

CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1000 mL
Catalog number H27080.AU
also known as H27080-AU
Price (EUR)
291,00
Each
Quantity:
1000 mL
Request bulk or custom format
Price (EUR)
291,00
Each
Chemical Identifiers
CAS7664-41-7
IUPAC Nameammonia
Molecular FormulaH3N
InChI KeyQGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILESN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (Aqueous acid-base Titration)1.90-2.20 M
Ammonia, 2M in methanol is used as a solvent in organic reactions and as a mobile phase in thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only