(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%
(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%
(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%
Thermo Scientific Chemicals

(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%

CAS: 56210-72-1 | C16H19N | 225.335 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27106.06
also known as H27106-06
Price (EUR)
152,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
152,00
Each
Chemical Identifiers
CAS56210-72-1
IUPAC Namebis(1-phenylethyl)amine
Molecular FormulaC16H19N
InChI KeyNXLACVVNHYIYJN-UHFFFAOYNA-N
SMILESCC(NC(C)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Optical Rotation-193° to -198° (Neat)
Refractive Index1.5510-1.5530 @ 20°C
(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhuang, Y.; He, Y.; Zhou, Z.; Xia, W.; Cheng, C.; Wang, M.; Chen, B.; Zhou, Z.; Pang, J.; Qiu, L. Synthesis of a Class of Chiral-Bridged Phosphoramidite Ligands and Their Applications in the First Iridium-Catalyzed Asymmetric Addition of Arylboronic Acids to Isatins. J. Org. Chem. 2015, 80 (14), 6968-6975.
  2. Dayaker, G.; Tilly, D.; Chevallier, F.; Hilmersson, G.; Gros, P. C.; Mongin, F. Enantioselective Metalation of N,N-Diisopropylferrocenecarboxamide and Methyl Ferrocenecarboxylate Using Lithium-Metal Chiral Bases. Eur. J. Org. Chem. 2012, 2012 (30), 6051-6057.