N-Boc-cis-4-fluoro-L-proline, 97%, Thermo Scientific Chemicals
N-Boc-cis-4-fluoro-L-proline, 97%, Thermo Scientific Chemicals
N-Boc-cis-4-fluoro-L-proline, 97%, Thermo Scientific Chemicals
N-Boc-cis-4-fluoro-L-proline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-cis-4-fluoro-L-proline, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H27225.03
also known as H27225-03
1 g
Catalog number H27225.03
also known as H27225-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS203866-13-1
IUPAC Name1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid
Molecular FormulaC10H16FNO4
InChI KeyYGWZXQOYEBWUTH-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CC(F)CC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only