4-Hydroxy-3,5-dimethoxyphenylacetic acid, 97%
4-Hydroxy-3,5-dimethoxyphenylacetic acid, 97%
4-Hydroxy-3,5-dimethoxyphenylacetic acid, 97%
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethoxyphenylacetic acid, 97%

CAS: 4385-56-2 | C10H12O5 | 212.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27231.03
also known as H27231-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS4385-56-2
IUPAC Name2-(4-hydroxy-3,5-dimethoxyphenyl)acetic acid
Molecular FormulaC10H12O5
InChI KeyBQBQKSSTFGCRQL-UHFFFAOYSA-N
SMILESCOC1=CC(CC(O)=O)=CC(OC)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Assay (Silylated GC)> 96.0%
Appearance (Color)White to cream or pale brown
Assay (Aqueous acid-base Titration)> 96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only