2-Bromoquinoline, 96%
2-Bromoquinoline, 96%
2-Bromoquinoline, 96%
Thermo Scientific Chemicals

2-Bromoquinoline, 96%

CAS: 2005-43-8 | C9H6BrN | 208.06 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
產品號碼 H27302.14
亦稱為 H27302-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS2005-43-8
規格Specification Sheet規格表
Appearance (Color)White to yellow to red
FormCrystals or powder or crystalline powder
Assay (GC)≥95.0%
Melting Point (clear melt)43.0-52.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only