5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%
Thermo Scientific Chemicals

5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%

CAS: 70978-54-0 | C8H6BrNO4 | 260.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27331.14
also known as H27331-14
Price (EUR)
113,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
113,00
Each
Add to cart
Chemical Identifiers
CAS70978-54-0
IUPAC Name1-(5-bromo-2-hydroxy-3-nitrophenyl)ethan-1-one
Molecular FormulaC8H6BrNO4
InChI KeyCLNIBJASCGZXHH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(Br)=CC(=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Pale yellow to yellow
Melting Point (clear melt)127.0-137.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only