2'-Aminoacetanilide, 98%
2'-Aminoacetanilide, 98%
2'-Aminoacetanilide, 98%
Thermo Scientific Chemicals

2'-Aminoacetanilide, 98%

CAS: 34801-09-7 | C8H10N2O | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27343.14
also known as H27343-14
Price (EUR)
195,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
195,00
Each
Chemical Identifiers
CAS34801-09-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale cream to cream
Melting Point (clear melt)131.0-137.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only