N,S-Dimethylisothiouronium hydriodide, 98%
N,S-Dimethylisothiouronium hydriodide, 98%
N,S-Dimethylisothiouronium hydriodide, 98%
N,S-Dimethylisothiouronium hydriodide, 98%
Thermo Scientific Chemicals

N,S-Dimethylisothiouronium hydriodide, 98%

CAS: 41306-45-0 | C3H9IN2S | 232.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27439.14
also known as H27439-14
Price (TWD)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS41306-45-0
IUPAC NameN'-methyl(methylsulfanyl)methanimidamide hydroiodide
Molecular FormulaC3H9IN2S
InChI KeyUDUMWDDMSHXGGQ-UHFFFAOYSA-N
SMILESI.CSC(N)=NC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Iodide)≥97.5 to ≤102.5%
Melting Point (clear melt)133-138?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only