Thermo Scientific Chemicals

(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific Chemicals

Catalog number: H27499.03
1 g, Each
Thermo Scientific Chemicals

(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific Chemicals

Catalog number: H27499.03
1 g, Each
Quantity
Catalog number: H27499.03
also known as H27499-03
Price (USD)
Price: 152.00
Online price: 131.65
Your price:
Quantity
-

Chemical Identifiers

CAS
195604-39-8
IUPAC Name
1-cyclopropylethan-1-amine
Molecular Formula
C5H11N
InChI Key
IXCXVGWKYIDNOS-UHFFFAOYNA-N
SMILES
CC(N)C1CC1
Assay (GC)
≥97.5%
Refractive Index
1.4245-1.4585 @ 20?C
Enantiomeric excess
≥98.0%
Optical Rotation
+27.5 ? 1.0? (neat)
Appearance (Color)
Clear colorless to pale yellow

Description

(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text