7-Bromo-2H-1,4-benzothiazin-3(4H)-one, 97%
7-Bromo-2H-1,4-benzothiazin-3(4H)-one, 97%
7-Bromo-2H-1,4-benzothiazin-3(4H)-one, 97%
Thermo Scientific Chemicals

7-Bromo-2H-1,4-benzothiazin-3(4H)-one, 97%

CAS: 90814-91-8 | C8H6BrNOS | 244.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27563.03
also known as H27563-03
Price (EUR)
104,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS90814-91-8
IUPAC Name7-bromo-3,4-dihydro-2H-1,4-benzothiazin-3-one
Molecular FormulaC8H6BrNOS
InChI KeyMDFPWMKBRDOSGB-UHFFFAOYSA-N
SMILESBrC1=CC2=C(NC(=O)CS2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white to cream
FormPowder
Assay (HPLC)≥96.0%
Melting Point (clear melt)209-215?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only