4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%
4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%
4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%
Thermo Scientific Chemicals

4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%

CAS: 172843-97-9 | C11H19NO5 | 245.275 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27578.MD
also known as H27578-MD
Price (EUR)
69,60
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
69,60
Each
Chemical Identifiers
CAS172843-97-9
IUPAC Name4-{[(tert-butoxy)carbonyl]amino}oxane-4-carboxylate
Molecular FormulaC11H18NO5
InChI KeySPPDKPRJPFTBEV-UHFFFAOYSA-M
SMILESCC(C)(C)OC(=O)NC1(CCOCC1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. (15 g/L) (25°C)

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Daniela Fattori.; Cristina Rossi.; Christopher I. Fincham.; Valerio Caciagli.; Fernando Catrambone.; Piero D'Andrea.; Patrizia Felicetti.; Martina Gensini.; Elena Marastoni.; Rossano Nannicini.; Marielle Paris.; Rosa Terracciano.; Alessandro Bressan.; Sandro Giuliani.; Carlo A. Maggi.; Stefania Meini .; Claudio Valenti.; Laura Quartara. Design and Synthesis of Novel Sulfonamide-Containing Bradykinin hB2 Receptor Antagonists. 2. Synthesis and Structure-Activity Relationships of α,α-Cycloalkylglycine Sulfonamides.J. Med. Chem. 2007, 50 (3), 550-565 .