Thermo Scientific Chemicals

Methyl 2-formylthiophene-4-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H27597.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-formylthiophene-4-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H27597.06
5 g, Each
Quantity
Catalog number: H27597.06
also known as H27597-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
67808-66-6
IUPAC Name
methyl 5-formylthiophene-3-carboxylate
Molecular Formula
C7H6O3S
InChI Key
GNXNZRYWBFMVHK-UHFFFAOYSA-N
SMILES
COC(=O)C1=CSC(C=O)=C1
Form
powder
Appearance (Color)
Brown
Assay (HPLC)
> 94.0%
Proton NMR
conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text