N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals
N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals
N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H27782.06
also known as H27782-06
5 g
Catalog number H27782.06
also known as H27782-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS102195-80-2
IUPAC Name1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate
Molecular FormulaC11H17NO5
InChI KeyUPBHYYJZVWZCOZ-UHFFFAOYNA-N
SMILESCOC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Melting Point (clear melt)41.0-47.0°C
Appearance (Color)White to yellow to pale brown
Assay (GC)≥96.0%
Optical Rotation+8° to +12° (c=1 in chloroform)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only