Thermo Scientific Chemicals

4-Phenyl-3-butyn-2-one, 96%, Thermo Scientific Chemicals

Catalog number: H27826.06
5 g, Each
Thermo Scientific Chemicals

4-Phenyl-3-butyn-2-one, 96%, Thermo Scientific Chemicals

Catalog number: H27826.06
5 g, Each
Quantity
Catalog number: H27826.06
also known as H27826-06
Price (USD)
Price: 212.00
Online price: 182.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1817-57-8
IUPAC Name
4-phenylbut-3-yn-2-one
Molecular Formula
C10H8O
InChI Key
UPEUQDJSUFHFQP-UHFFFAOYSA-N
SMILES
CC(=O)C#CC1=CC=CC=C1
Form
Liquid
Assay (GC)
≥95.0%
Refractive Index
1.5710-1.5770 @ 20?C
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text