Trimethoxy(pentamethylcyclopentadienyl)titanium(IV), 97+%
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV), 97+%
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV), 97+%
Thermo Scientific Chemicals

Trimethoxy(pentamethylcyclopentadienyl)titanium(IV), 97+%

CAS: 123927-75-3 | C13H24O3Ti+ | 276.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27885.03
also known as H27885-03
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS123927-75-3
Molecular FormulaC13H24O3Ti
InChI KeyGQQYDESAAOEXPK-UHFFFAOYSA-N
SMILES[Ti+4].C[O-].C[O-].C[O-].C[C]1[C](C)[C](C)[C](C)[C]1C
Molecular Weight (g/mol)276.20
SpecificationsSpecification SheetSpecification Sheet
Assay by NMR≥97.0% (by H-NMR)
Proton NMRConforms to structure
Appearance (Color)Pale yellow to yellow
Impurity contentChloride Cl: ≤50ppm
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only