6-Chloro-3-pyridineacetic acid, 97%
6-Chloro-3-pyridineacetic acid, 97%
6-Chloro-3-pyridineacetic acid, 97%
6-Chloro-3-pyridineacetic acid, 97%
Thermo Scientific Chemicals

6-Chloro-3-pyridineacetic acid, 97%

CAS: 39891-13-9 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
50 g
產品號碼 H27966.09
亦稱為 H27966-09
價格 (TWD)
-
申請報價
Quantity:
10 g
Request bulk or custom format
化學識別
CAS39891-13-9
IUPAC Name2-(6-chloropyridin-3-yl)acetic acid
Molecular FormulaC7H6ClNO2
InChI KeyXWVPSJPQWOVRHJ-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(Cl)N=C1
檢視更多
規格Specification Sheet規格表
FormPowder
Assay (HPLC)≥96.0%
Appearance (Color)Pale cream to cream to brown
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only