3-Iodobenzeneboronic acid pinacol ester, 97%
3-Iodobenzeneboronic acid pinacol ester, 97%
3-Iodobenzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-Iodobenzeneboronic acid pinacol ester, 97%

CAS: 408492-28-4 | C12H16BIO2 | 329.97 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 H28115.06
亦稱為 H28115-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS408492-28-4
IUPAC Name2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC12H16BIO2
InChI KeyHSHFNMSHDHAHDN-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1
檢視更多
規格Specification Sheet規格表
FormPowder
Appearance (Color)Cream
Assay (GC)≥96.0%
Melting Point (clear melt)67.0-76.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only