It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Es una materia prima importante y se utiliza como intermediario en la síntesis orgánica, productos farmacéuticos, productos agroquímicos y colorantes.
Solubilidad
Soluble en agua.
Notas
Debe almacenarse en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de agentes oxidantes.
RUO – Research Use Only
General References:
- Alenka Paquet. Introduction of 9-fluorenylmethyloxycarbonyl, trichloroethoxycarbonyl, and benzyloxycarbonyl amine protecting groups into O-unprotected hydroxyamino acids using succinimidyl carbonates. Canadian Journal of Chemistry. 1982, 60(8), 976-980.
- Yuo-Ling Chou; Michael M. Morrissey; Raju Mohan. Novel serine-based linker for the solid-phase synthesis of organic compounds.Tetrahedron Letters. 1998, 39(8), 757-760.