4-(Boc-amino)benzeneboronic acid, 97%
4-(Boc-amino)benzeneboronic acid, 97%
4-(Boc-amino)benzeneboronic acid, 97%
Thermo Scientific Chemicals

4-(Boc-amino)benzeneboronic acid, 97%

CAS: 380430-49-9 | C11H16BNO4 | 237.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28799.06
also known as H28799-06
Price (EUR)
123,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
123,00
Each
Chemical Identifiers
CAS380430-49-9
IUPAC Name(4-{[(tert-butoxy)carbonyl]amino}phenyl)boronic acid
Molecular FormulaC11H16BNO4
InChI KeyUBVOLHQIEQVXGM-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to cream
FormPowder
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only