2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
Thermo Scientific Chemicals

2-Cyanobenzaldehyde, 98%

CAS: 7468-67-9 | C8H5NO | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H29027.03
also known as H29027-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7468-67-9
IUPAC Name2-formylbenzonitrile
Molecular FormulaC8H5NO
InChI KeyQVTPWONEVZJCCS-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to orange or pale brown
FormCrystalline powder or powder or crystals and/or lumpy powder/chunks
Assay (GC)≥96.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only