5-Formylthiophene-2-boronic acid, 97%
5-Formylthiophene-2-boronic acid, 97%
5-Formylthiophene-2-boronic acid, 97%
Thermo Scientific Chemicals

5-Formylthiophene-2-boronic acid, 97%

CAS: 4347-33-5 | C5H5BO3S | 155.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H29289.03
also known as H29289-03
Price (EUR)
83,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
83,20
Each
Chemical Identifiers
CAS4347-33-5
IUPAC Name(5-formylthiophen-2-yl)boronic acid
Molecular FormulaC5H5BO3S
InChI KeyDEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥89.0%
Proton NMRConforms to structure
FormPowder
Assay (HPLC)≥96.0%
Appearance (Color)White to cream or orange to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only