1-Adamantanamine, 98%
1-Adamantanamine, 98%
1-Adamantanamine, 98%
Thermo Scientific Chemicals

1-Adamantanamine, 98%

CAS: 768-94-5 | C10H17N | 151.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H30076.22
also known as H30076-22
Price (EUR)
549,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
549,00
Each
Chemical Identifiers
CAS768-94-5
IUPAC Nameadamantan-1-amine
Molecular FormulaC10H17N
InChI KeyDKNWSYNQZKUICI-UHFFFAOYSA-N
SMILESNC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder.
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. Yang, H.; Liu, Y.; Yang, L.; Liu, K.; Wang, Z.; Zhang, X. Cucurbit[7]uril as a protective agent: controlling photochemistry and detecting 1-adamantanamine. Chem. Commun. 2013, 49, 3905-3907.
  2. Bose, A.; Das, J. K.; Das, N. Pore modification of deca-dodecasil-rhombohedral zeolite membrane by carbon loading from in situ decomposition of 1-adamantanamine for improved gas separation. RSC Adv. 2015, 5, , 67195-67205.