2-Bromothiazole-5-carboxaldehyde, 95%, Thermo Scientific Chemicals
2-Bromothiazole-5-carboxaldehyde, 95%, Thermo Scientific Chemicals
2-Bromothiazole-5-carboxaldehyde, 95%, Thermo Scientific Chemicals
2-Bromothiazole-5-carboxaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromothiazole-5-carboxaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H31096.03
also known as H31096-03
1 g
Catalog number H31096.03
also known as H31096-03
Price (EUR)
69,70
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
69,70
Each
Add to cart
Chemical Identifiers
CAS464192-28-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormPowder
Assay (GC)≥94.0%
Free acid (titration)≤2.5%
Melting Point (clear melt)103.0-115.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only