4-Fluoro-alpha-toluenesulfonyl chloride, 97%
4-Fluoro-alpha-toluenesulfonyl chloride, 97%
4-Fluoro-alpha-toluenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Fluoro-alpha-toluenesulfonyl chloride, 97%

CAS: 103360-04-9 | C7H6ClFO2S | 208.631 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mg
1 g
產品號碼 H31605.03
亦稱為 H31605-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS103360-04-9
IUPAC Name(4-fluorophenyl)methanesulfonyl chloride
Molecular FormulaC7H6ClFO2S
InChI KeyUUQGWVIRPCRTSA-UHFFFAOYSA-N
SMILESFC1=CC=C(CS(Cl)(=O)=O)C=C1
檢視更多
規格Specification Sheet規格表
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)53.0-63.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only