4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%
4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%
4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%
Thermo Scientific Chemicals

4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%

CAS: 124811-71-8 | C8H4F3NO | 187.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31639.03
also known as H31639-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS124811-71-8
IUPAC Name4-hydroxy-3-(trifluoromethyl)benzonitrile
Molecular FormulaC8H4F3NO
InChI KeyQXIFYONUKBXFTH-UHFFFAOYSA-N
SMILESOC1=C(C=C(C=C1)C#N)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)170-176?C
Appearance (Color)Cream to brown or yellow
Assay (GC)≥98.0%
FormPowder
As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Wenna Huang,; Lidan Xing,; Yating Wanga, Mengqing Xua, Weishan Lia,; Fengchao Xieb,; Shengan Xia. 4-(Trifluoromethyl)-benzonitrile: A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery . Journal of Power Sources. 2014, 267 560-565.
  2. authors. title . Journal name. year of publication , volume issue , pg no.